CH4424845352 ChemBridge Corporation

(2-amino-4,5-dihydro-1,3-thiazol-5-yl)methanol hydrobromide

  • MDL number MFCD07773076
  • Cas Number : 35525-88-3
  • Molecular Weight : 213.0961

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004234

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004234
Availability details for
Quantity
  • Availability
  • Add to Cart