CH4424845402 ChemBridge Corporation

4-(3-methylphenoxy)piperidine hydrochloride

  • MDL number MFCD09064908
  • Cas Number : 65367-96-6
  • Molecular Weight : 227.7304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004488

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4004488
Availability details for
Quantity
  • Availability
  • Add to Cart