CH4424845498 ChemBridge Corporation

[3-(2-methyl-1H-imidazol-1-yl)propyl]amine dihydrochloride

  • MDL number MFCD06801042
  • Cas Number : 1059626-17-3
  • Molecular Weight : 212.1201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4005658

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4005658
Availability details for
Quantity
  • Availability
  • Add to Cart