CH4424845510 ChemBridge Corporation

(4,5,6,7-tetrahydro-1,3-benzothiazol-2-ylmethyl)amine dihydrochloride

  • MDL number MFCD08689810
  • Cas Number : 1240528-53-3
  • Molecular Weight : 241.1812

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4005754

Safety Information

MSDS https://www.hit2lead.com/sds/4005754_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart