CH4424845651 ChemBridge Corporation

4-piperidinecarbonitrile hydrochloride

  • MDL number MFCD09037930
  • Cas Number : 240401-22-3
  • Molecular Weight : 146.6179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009819

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/4009819
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart