CH4424845743 ChemBridge Corporation

1-phenyl-4-piperidinamine dihydrochloride

  • MDL number MFCD23135281
  • Cas Number : 1082662-38-1
  • Molecular Weight : 249.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010828

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010828
Availability details for
Quantity
  • Availability
  • Add to Cart