CH4424845802 ChemBridge Corporation

3-amino-4-methyl-1,3-dihydro-2H-indol-2-one hydrochloride

  • MDL number MFCD11502899
  • Cas Number : 1214103-58-8
  • Molecular Weight : 198.6494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011502

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4011502
Availability details for
Quantity
  • Availability
  • Add to Cart