CH4424845810 ChemBridge Corporation

methyl (3-methyl-2-oxo-1-piperazinyl)acetate hydrobromide

  • MDL number MFCD13193922
  • Cas Number : 1609396-06-6
  • Molecular Weight : 267.1203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011526

Safety Information

MSDS https://www.hit2lead.com/sds/4011526_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart