CH4424845812 ChemBridge Corporation

1-(3-methoxyphenyl)-3-methyl-2-piperazinone hydrochloride

  • MDL number MFCD13186166
  • Cas Number : 1609395-39-2
  • Molecular Weight : 256.7286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011537

Safety Information

MSDS https://www.hit2lead.com/sds/4011537_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart