CH4424845814 ChemBridge Corporation

1-benzyl-3-methyl-2-piperazinone hydrobromide

  • MDL number MFCD11841260
  • Cas Number : 1214717-49-3
  • Molecular Weight : 285.1802

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011539

Safety Information

MSDS https://www.hit2lead.com/sds/4011539_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart