CH4424845836 ChemBridge Corporation

[(1-acetyl-3-piperidinyl)methyl]amine hydrochloride

  • MDL number MFCD11841262
  • Cas Number : 943843-70-7
  • Molecular Weight : 192.6864

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011882

Safety Information

MSDS https://www.hit2lead.com/sds/4011882_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart