CH4424845841 ChemBridge Corporation

{[2-(1-pyrrolidinyl)-1,3-thiazol-4-yl]methyl}amine dihydrochloride

  • MDL number MFCD13857383
  • Cas Number : 1211465-93-8
  • Molecular Weight : 256.1958

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012024

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4012024
Availability details for
Quantity
  • Availability
  • Add to Cart