CH4424845842 ChemBridge Corporation

{[2-(4-morpholinyl)-1,3-thiazol-4-yl]methyl}amine dihydrochloride

  • MDL number MFCD13857382
  • Cas Number : 1211479-11-6
  • Molecular Weight : 272.1952

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012025

Safety Information

MSDS https://www.hit2lead.com/sds/4012025_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart