CH4424845847 ChemBridge Corporation

{[3-(2-methoxyethyl)-1,2,4-oxadiazol-5-yl]methyl}methylamine trifluoroacetate

  • MDL number MFCD13193844
  • Cas Number : 1609400-25-0
  • Molecular Weight : 285.2204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012064

Safety Information

MSDS https://www.hit2lead.com/sds/4012064_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart