CH4424845862 ChemBridge Corporation

3-isopropyl-5-(2-pyrrolidinyl)-1,2,4-oxadiazole trifluoroacetate

  • MDL number MFCD26959467
  • Cas Number : 1609406-28-1
  • Molecular Weight : 295.2582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3COOH
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012107

Safety Information

MSDS https://www.hit2lead.com/sds/4012107_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart