CH4424845866 ChemBridge Corporation

3-(3-isopropyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride

  • MDL number MFCD12028440
  • Cas Number : 1185296-16-5
  • Molecular Weight : 231.7224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012132

Safety Information

MSDS https://www.hit2lead.com/sds/4012132_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart