CH4424845868 ChemBridge Corporation

3-(3-phenyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride

  • MDL number MFCD12028442
  • Cas Number : 1185302-70-8
  • Molecular Weight : 265.7386

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012137

Safety Information

MSDS https://www.hit2lead.com/sds/4012137_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart