CH4424845938 ChemBridge Corporation

2-(2-methoxyphenyl)ethanimidamide hydrochloride

  • MDL number MFCD11841268
  • Cas Number : 1029718-90-8
  • Molecular Weight : 200.6653

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012625

Safety Information

MSDS https://www.hit2lead.com/sds/4012625_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart