CH4424845964 ChemBridge Corporation

{[5-(2-furyl)-1,2,4-oxadiazol-3-yl]methyl}amine hydrochloride

  • MDL number MFCD26959704
  • Cas Number : 1609400-88-5
  • Molecular Weight : 201.6103

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013177

Safety Information

MSDS https://www.hit2lead.com/sds/4013177_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart