CH4424845971 ChemBridge Corporation

isoxazolidine hydrochloride

  • MDL number MFCD11100465
  • Cas Number : 39657-45-9
  • Molecular Weight : 109.5547

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013261

Safety Information

MSDS https://www.hit2lead.com/sds/4013261_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart