CH4424845999 ChemBridge Corporation

(2-methyl-1-benzofuran-5-yl)amine hydrochloride

  • MDL number MFCD00035194
  • Cas Number : 23968-37-8
  • Molecular Weight : 183.6348

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013733

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4013733
Availability details for
Quantity
  • Availability
  • Add to Cart