CH4424846028 ChemBridge Corporation

4-amino-1-(2-phenylethyl)-2-pyrrolidinone hydrochloride

  • MDL number MFCD11506523
  • Cas Number : 1177362-59-2
  • Molecular Weight : 240.7292

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014090

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4014090
Availability details for
Quantity
  • Availability
  • Add to Cart