CH4424846031 ChemBridge Corporation

4-amino-1-(3-methoxyphenyl)-2-pyrrolidinone hydrochloride

  • MDL number MFCD11506524
  • Cas Number : 1177302-83-8
  • Molecular Weight : 242.702

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014096

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4014096
Availability details for
Quantity
  • Availability
  • Add to Cart