CH4424846032 ChemBridge Corporation

4-amino-1-(2-methoxyphenyl)-2-pyrrolidinone hydrochloride

  • MDL number MFCD11506525
  • Cas Number : 1177283-77-0
  • Molecular Weight : 242.702

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014097

Safety Information

MSDS https://www.hit2lead.com/sds/4014097_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart