CH4424846072 ChemBridge Corporation

[2-(aminomethyl)benzyl]dimethylamine dihydrochloride

  • MDL number MFCD11841287
  • Cas Number : 53369-78-1
  • Molecular Weight : 237.1693

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014713

Safety Information

MSDS https://www.hit2lead.com/sds/4014713_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart