CH4424846080 ChemBridge Corporation

[(1-benzyl-1H-imidazol-2-yl)methyl]methylamine dihydrochloride

  • MDL number MFCD18071264
  • Cas Number : 1189471-03-1
  • Molecular Weight : 274.1895

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014751

Safety Information

MSDS https://www.hit2lead.com/sds/4014751_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart