CH4424846085 ChemBridge Corporation

N-[(5-cyclopropyl-1,2,4-oxadiazol-3-yl)methyl]ethanamine hydrochloride

  • MDL number MFCD13186095
  • Cas Number : 1257850-59-1
  • Molecular Weight : 203.6693

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014788

Safety Information

MSDS https://www.hit2lead.com/sds/4014788_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart