CH4424846131 ChemBridge Corporation

2-aminoisonicotinic acid hydrochloride

  • MDL number MFCD09870077
  • Cas Number : 1185560-43-3
  • Molecular Weight : 174.585

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4015121

Safety Information

MSDS https://www.hit2lead.com/sds/4015121_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart