CH4424846196 ChemBridge Corporation

{1-[4-(3-methoxypropyl)-4H-1,2,4-triazol-3-yl]ethyl}amine dihydrochloride

  • MDL number MFCD13186215
  • Cas Number : 1609407-91-1
  • Molecular Weight : 257.1607

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016279

Safety Information

MSDS https://www.hit2lead.com/sds/4016279_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart