CH4424846211 ChemBridge Corporation

amino(1,3,5-trimethyl-1H-pyrazol-4-yl)acetic acid dihydrochloride hydrate

  • MDL number MFCD26959535
  • Cas Number : 1025555-34-3
  • Molecular Weight : 274.1449

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016615

Safety Information

MSDS https://www.hit2lead.com/sds/4016615_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart