CH4424846214 ChemBridge Corporation

1,4-dimethyl-4-piperidinecarboxylic acid hydrochloride

  • MDL number MFCD11841301
  • Cas Number : 1185325-87-4
  • Molecular Weight : 193.6711

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016670

Safety Information

MSDS https://www.hit2lead.com/sds/4016670_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart