CH4424846219 ChemBridge Corporation

[3-(4-methoxyphenyl)propyl]amine hydrochloride

  • MDL number MFCD09991708
  • Cas Number : 36397-51-0
  • Molecular Weight : 201.6931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016880

Safety Information

MSDS https://www.hit2lead.com/sds/4016880_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart