CH4424846228 ChemBridge Corporation

4-(4-methylphenyl)-1-piperazinecarboximidamide sulfate hydrate

  • MDL number MFCD13193872
  • Molecular Weight : 334.3919

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 0.5H2SO4 H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017015

Safety Information

MSDS https://www.hit2lead.com/sds/4017015_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart