CH4424846230 ChemBridge Corporation

4-phenyl-1-piperazinecarboximidamide sulfate hydrate

  • MDL number MFCD01748424
  • Cas Number : 17238-58-3
  • Molecular Weight : 320.3653

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 0.5H2SO4 H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017018

Safety Information

MSDS https://www.hit2lead.com/sds/4017018_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart