CH4424846247 ChemBridge Corporation

N-(1,3-thiazol-2-ylmethyl)cyclopropanamine dihydrochloride

  • MDL number MFCD11841305
  • Cas Number : 1185383-89-4
  • Molecular Weight : 227.1546

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017446

Safety Information

MSDS https://www.hit2lead.com/sds/4017446_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart