CH4424846251 ChemBridge Corporation

N-ethyl-2,2-dimethyl-1-propanamine hydrochloride

  • MDL number MFCD11226476
  • Cas Number : 22675-77-0
  • Molecular Weight : 151.6775

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017478

Safety Information

MSDS https://www.hit2lead.com/sds/4017478_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart