CH4424846265 ChemBridge Corporation

3-(1-azocanyl)propanoic acid hydrochloride

  • MDL number MFCD12026859
  • Cas Number : 1185294-68-1
  • Molecular Weight : 221.7243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017704

Safety Information

MSDS https://www.hit2lead.com/sds/4017704_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart