CH4424846294 ChemBridge Corporation

4-(4-morpholinyl)butanoic acid hydrochloride

  • MDL number MFCD06801119
  • Cas Number : 39493-84-0
  • Molecular Weight : 209.6705

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4018164

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4018164
Availability details for
Quantity
  • Availability
  • Add to Cart