CH4424846295 ChemBridge Corporation

[3-chloro-2-(1-piperidinyl)phenyl]amine dihydrochloride

  • MDL number MFCD13186183
  • Cas Number : 1443278-97-4
  • Molecular Weight : 283.6251

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012421

Safety Information

MSDS https://www.hit2lead.com/sds/4012421_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart