CH4424846325 ChemBridge Corporation

4-(methylthio)benzenecarboximidamide hydrochloride

  • MDL number MFCD20502058
  • Cas Number : 860685-96-7
  • Molecular Weight : 202.7043

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4019315

Safety Information

MSDS https://www.hit2lead.com/sds/4019315_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart