CH4424846354 ChemBridge Corporation

1-(2-pyrimidinyl)-4-piperidinamine dihydrochloride

  • MDL number MFCD11052947
  • Cas Number : 1179369-48-2
  • Molecular Weight : 251.1561

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4020038

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4020038
Availability details for
Quantity
  • Availability
  • Add to Cart