CH4424846399 ChemBridge Corporation

(1-benzyl-4-piperidinyl)acetic acid hydrochloride

  • MDL number MFCD13250102
  • Cas Number : 99944-02-2
  • Molecular Weight : 269.7671

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4021953

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4021953
Availability details for
Quantity
  • Availability
  • Add to Cart