CH4424846418 ChemBridge Corporation

(3-isopropyl-4-methylphenyl)amine hydrochloride

  • MDL number MFCD13186094
  • Cas Number : 91251-36-4
  • Molecular Weight : 185.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4022447

Safety Information

MSDS https://www.hit2lead.com/sds/4022447_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart