CH4424846456 ChemBridge Corporation

2-methoxybutanohydrazide hydrochloride

  • MDL number MFCD08559375
  • Cas Number : 1049749-93-0
  • Molecular Weight : 168.6219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024078

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4024078
Availability details for
Quantity
  • Availability
  • Add to Cart