CH4424846457 ChemBridge Corporation

2-isopropoxybutanohydrazide hydrochloride

  • MDL number MFCD08727982
  • Cas Number : 1049749-98-5
  • Molecular Weight : 196.6751

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024079

Safety Information

MSDS https://www.hit2lead.com/sds/4024079_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart