CH4424846475 ChemBridge Corporation

N-(4-methylbenzyl)-1-butanamine hydrochloride

  • MDL number MFCD07105374
  • Cas Number : 16183-22-5
  • Molecular Weight : 213.7469

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024842

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4024842
Availability details for
Quantity
  • Availability
  • Add to Cart