CH4424846488 ChemBridge Corporation

N-butylcyclopentanamine hydrochloride

  • MDL number MFCD08559402
  • Cas Number : 1049750-21-1
  • Molecular Weight : 177.7148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024875

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4024875
Availability details for
Quantity
  • Availability
  • Add to Cart