CH4424846491 ChemBridge Corporation

N-(2-methylbenzyl)-2-butanamine hydrochloride

  • MDL number MFCD07110300
  • Cas Number : 1049750-39-1
  • Molecular Weight : 213.7469

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024882

Safety Information

MSDS https://www.hit2lead.com/sds/4024882_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart