CH4424846502 ChemBridge Corporation

N-(4-chlorobenzyl)ethanamine hydrochloride

  • MDL number MFCD07106854
  • Cas Number : 102236-18-0
  • Molecular Weight : 206.1122

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4024922

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4024922
Availability details for
Quantity
  • Availability
  • Add to Cart