CH4424846585 ChemBridge Corporation

(4-morpholin-4-ylbenzyl)amine hydrochloride

  • MDL number MFCD25955516
  • Cas Number : 1106986-47-3
  • Molecular Weight : 228.7185

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012455

Safety Information

MSDS https://www.hit2lead.com/sds/4012455_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart